Learn More
Medchemexpress LLC Maleimide | 541-59-3 | 97.07 | 5 G
SDP
Catalog No. 5000460399
Encompass_Preferred
Maleimide can be used for the production of antibody-drug conjugate (ADC) in cancer research. It can also be leveraged for the preparation of fluorogenic probes, which are mainly used for the specific detection of thiol analytes.
  • Used in the production of antibody-drug conjugates (ADCs) for cancer research.
  • Can be used for the preparation of fluorogenic probes for specific detection of thiol analytes.
  • Binds rapidly and covalently to the sulfhydryl (-SH) group of cysteine residues.
  • Can be used in drug delivery systems for improved cell uptake efficiency.
Catalog No. 50-004-60399 Supplier Medchemexpress LLC Supplier No. HYW0073245G
Add to Cart
Add to Cart
Maleimide can be used for the production of antibody-drug conjugate (ADC) in cancer research. It can also be leveraged for the preparation of fluorogenic probes, which are mainly used for the specific detection of thiol analytes.
  • Used in the production of antibody-drug conjugates (ADCs) for cancer research.
  • Can be used for the preparation of fluorogenic probes for specific detection of thiol analytes.
  • Binds rapidly and covalently to the sulfhydryl (-SH) group of cysteine residues.
  • Can be used in drug delivery systems for improved cell uptake efficiency.