Learn More
Selleck Chemical LLC Maleimide S3141-100mg
SDP
Catalog No. 503112986
Encompass_Preferred
Maleimide exhibits fluorescence quenching ability and can be used for the specific detection of thiol analytes as fluorogenic probes Maleimide is also used for production of antibody-drug conjugate (ADC) which is used in cancer research
Catalog No. 50-311-2986 Supplier Selleck Chemical LLC Supplier No. S3141100MG
Maleimide exhibits fluorescence quenching ability and can be used for the specific detection of thiol analytes as fluorogenic probes Maleimide is also used for production of antibody-drug conjugate (ADC) which is used in cancer research