Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Maleimide Activated R-Phycoerythrin
SDP

Catalog No. 4030002
Change view
Click to view available options
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
40-300-02 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-300-02 Supplier Novus Biologicals™ Supplier No. 4030002

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen

The reactive maleimide group that has been added to the R-Phycoerythrin allows conjugation reactions to be performed very efficiently at physiological pH.

Dilutions
  • Immunohistochemistry 1:10-1:500
  • Immunohistochemistry- Frozen
  • Immunohistochemistry- Paraffin

Recommended Storage

Store at -20.Avoid freeze-thaw cycles.

Specifications

Content And Storage Store at -20.Avoid freeze-thaw cycles.
Type Maleimide Activated R-Phycoerythrin
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.