Learn More
Medchemexpress LLC Maleimide-C10-NHS ester | 87981-04-2 | 98.0% | 378.42 | 100 MG
SDP
Catalog No. 5000453118
Encompass_Preferred
Maleimide-C10-NHS ester is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. These compounds leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for selective target protein degradation
Catalog No. 50-004-53118 Supplier Medchemexpress LLC Supplier No. HYW009037100MG
Maleimide-C10-NHS ester is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. These compounds leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for selective target protein degradation