Learn More
Medchemexpress LLC Maleimide-C10-NHS ester | 87981-04-2 | 98.0% | 378.42 | 250 MG
SDP
Catalog No. 5000451520
Encompass_Preferred
Maleimide-C10-NHS ester is identified as an alkyl/ether-based PROTAC linker, playing a role in the synthesis of PROTACs. PROTACs themselves are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. These molecules are designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation.
  • Alkyl/ether-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • PROTACs exploit the intracellular ubiquitin-proteasome system.
  • Enables selective degradation of target proteins.
Catalog No. 50-004-51520 Supplier Medchemexpress LLC Supplier No. 250MGHYW009037
Maleimide-C10-NHS ester is identified as an alkyl/ether-based PROTAC linker, playing a role in the synthesis of PROTACs. PROTACs themselves are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. These molecules are designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation.
  • Alkyl/ether-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • PROTACs exploit the intracellular ubiquitin-proteasome system.
  • Enables selective degradation of target proteins.