Learn More
Medchemexpress LLC Maleimide-DOTA | 1006711-90-5 | 99.6% | 526.54 | C22H34N6O9 | 25 MG
SDP
Catalog No. 5000203674
Encompass_Preferred
Maleimide-DOTA is a non-cleavable antibody-drug conjugate (ADC) linker that couples payloads to antibodies via maleimide chemistry for thiol-selective conjugation. Supplied for research use, it contains a DOTA chelator enabling radiolabeling and bioconjugation applications.
  • Non-cleavable maleimide-based linker.
  • DOTA chelator suitable for radiolabeling and metal coordination.
  • High purity (approximately 99.6%).
  • Soluble in DMSO and water; sonication recommended for dissolution.
  • Examples of in vivo vehicle protocols provided by manufacturer for formulation.
  • Store product at -20°C; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-03674 Supplier Medchemexpress LLC Supplier No. HY13354025MG
Maleimide-DOTA is a non-cleavable antibody-drug conjugate (ADC) linker that couples payloads to antibodies via maleimide chemistry for thiol-selective conjugation. Supplied for research use, it contains a DOTA chelator enabling radiolabeling and bioconjugation applications.
  • Non-cleavable maleimide-based linker.
  • DOTA chelator suitable for radiolabeling and metal coordination.
  • High purity (approximately 99.6%).
  • Soluble in DMSO and water; sonication recommended for dissolution.
  • Examples of in vivo vehicle protocols provided by manufacturer for formulation.
  • Store product at -20°C; in solvent, store at -80°C for long-term stability.