Learn More
Medchemexpress LLC Maleimide-PEG2 N-hydroxysuccinimide ester | 329364-72-9 | MFCD11041137 | 95.0% | 340.29 g/mol | C14H16N2O8 | 5 MG
SDP
Catalog No. 5000222659
Encompass_Preferred
Mal-PEG2-NHS is a PEG-based bifunctional linker combining a maleimide reactive group with an N-hydroxysuccinimide (NHS) ester. It enables selective conjugation between thiol and amine functional groups and is commonly used in PROTAC synthesis, bioconjugation, and antibody-drug conjugate linker construction. Supplied as a research-grade reagent for laboratory-scale synthesis and conjugation workflows.
  • Enables selective thiol-to-amine conjugation via maleimide and NHS chemistries.
  • Two-unit PEG spacer improves water solubility and reduces steric hindrance.
  • Suitable for PROTAC synthesis, bioconjugation, and ADC linker design.
  • Provided as a small-scale research reagent compatible with common solvents and buffers.
  • Characterized by analytical methods and typically supplied with a certificate of analysis.
Catalog No. 50-002-22659 Supplier Medchemexpress LLC Supplier No. HY1384305MG
Mal-PEG2-NHS is a PEG-based bifunctional linker combining a maleimide reactive group with an N-hydroxysuccinimide (NHS) ester. It enables selective conjugation between thiol and amine functional groups and is commonly used in PROTAC synthesis, bioconjugation, and antibody-drug conjugate linker construction. Supplied as a research-grade reagent for laboratory-scale synthesis and conjugation workflows.
  • Enables selective thiol-to-amine conjugation via maleimide and NHS chemistries.
  • Two-unit PEG spacer improves water solubility and reduces steric hindrance.
  • Suitable for PROTAC synthesis, bioconjugation, and ADC linker design.
  • Provided as a small-scale research reagent compatible with common solvents and buffers.
  • Characterized by analytical methods and typically supplied with a certificate of analysis.