Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Maleimide-Peg6-Succinimidyl Ester

Catalog No. 746193250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-619-3250MG 250 mg
74-619-350MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-619-3250MG Supplier Sigma Aldrich Supplier No. 746193250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Heterobifunctional crosslinker with an ethylene oxide spacer for linking amine- to sulfhydryl-containing compounds or biomolecules. Maleimide functional group will react with sulfhydryls and the succinimidyl ester group will react with amines. Spacer length is 31.7 angstroms.

Specifications

CAS 1137109-21-7
Molecular Formula C26H39N3O13
Linear Formula C26H39N3O13
MDL Number MFCD11041093
Quantity 250 mg
Synonym 25-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)-23-oxo-4,7,10,13,16,19-hexaoxa-22-azapentacosanoic acid 2,5-dioxo-1-pyrrolidinyl ester; Maleimide-PEG6-NHS ester
Molecular Weight (g/mol) 601.6 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.