Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Maleimido-tri(ethylene glycol)-propionic acid | 518044-40-1 | 98.0% | 1 G
SDP

Catalog No. 5000338643
Encompass_Preferred

Maleimido-tri(ethylene glycol)-propionic acid (Mal-PEG3-acid) is a cleavable ADC linker for synthesizing antibody-drug conjugates (ADCs). It facilitates site-specific cysteine mutant trastuzumab-A114C conjugation for neolymphostin-based ADC precursors and functions as a PEG-based PROTAC linker for PROTACs.

  • Cleavable ADC linker
  • PEG-based PROTAC linker
  • Enables the synthesis of antibody-drug conjugates (ADCs)
  • Facilitates site-specific conjugation for neolymphostin-based ADC precursors
  • Can be used in the synthesis of PROTACs
  • Appearance: colorless to light yellow liquid
  • Molecular weight: 301.29
  • Molecular formula: C13H19NO7
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-38643 Supplier Medchemexpress LLC Supplier No. HY1304261G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maleimido-tri(ethylene glycol)-propionic acid (Mal-PEG3-acid) is a cleavable ADC linker for synthesizing antibody-drug conjugates (ADCs). It facilitates site-specific cysteine mutant trastuzumab-A114C conjugation for neolymphostin-based ADC precursors and functions as a PEG-based PROTAC linker for PROTACs.

  • Cleavable ADC linker
  • PEG-based PROTAC linker
  • Enables the synthesis of antibody-drug conjugates (ADCs)
  • Facilitates site-specific conjugation for neolymphostin-based ADC precursors
  • Can be used in the synthesis of PROTACs
  • Appearance: colorless to light yellow liquid
  • Molecular weight: 301.29
  • Molecular formula: C13H19NO7
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.