Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Maleimido-tri(ethylene glycol)-propionic acid | 518044-40-1 | 98.0% | 301.29 | C13H19NO7 | 100 MG
SDP

Catalog No. 5000203717
Encompass_Preferred

Maleimido-tri(ethylene glycol)-propionic acid is a PEG-based maleimide linker used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PROTAC linker. It has formula C13H19NO7, molecular weight 301.29 g·mol⁻¹, and is supplied at 98.0% purity. Recommended storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solution -80°C (6 months) or -20°C (1 month).

  • Cleavable maleimide PEG3 linker for site-specific cysteine conjugation.
  • Suitable for ADC and PROTAC synthesis.
  • High purity, 98.0%.
  • Molecular weight 301.29 g·mol⁻¹.
  • CAS number 518044-40-1.
  • Available in quantities from milligram to gram scale, including 100 mg.
  • Store frozen or refrigerated according to storage guidance.

Catalog No. 50-002-03717 Supplier Medchemexpress LLC Supplier No. HY130426100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maleimido-tri(ethylene glycol)-propionic acid is a PEG-based maleimide linker used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PROTAC linker. It has formula C13H19NO7, molecular weight 301.29 g·mol⁻¹, and is supplied at 98.0% purity. Recommended storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solution -80°C (6 months) or -20°C (1 month).

  • Cleavable maleimide PEG3 linker for site-specific cysteine conjugation.
  • Suitable for ADC and PROTAC synthesis.
  • High purity, 98.0%.
  • Molecular weight 301.29 g·mol⁻¹.
  • CAS number 518044-40-1.
  • Available in quantities from milligram to gram scale, including 100 mg.
  • Store frozen or refrigerated according to storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.