Learn More
Medchemexpress LLC Maleimido-tri(ethylene glycol)-propionic acid | 518044-40-1 | 98.0% | 5 G
SDP
Catalog No. 5000338156
Encompass_Preferred
Maleimido-tri(ethylene glycol)-propionic acid is a cleavable linker utilized in the creation of antibody-drug conjugates (ADCs) and as a PEG-based PROTAC linker for the synthesis of PROTACs. It plays a role in preparing ADC precursors for specific cysteine mutant conjugation.
  • Functions as a cleavable linker.
  • Can be used to synthesize linker-payloads for conjugation.
  • Applicable in the preparation of ADCs.
  • Useful as a PEG-based linker for PROTAC synthesis.
  • Aids in creating neolymphostin-based ADC precursors.
Catalog No. 50-003-38156 Supplier Medchemexpress LLC Supplier No. HY1304265G
Maleimido-tri(ethylene glycol)-propionic acid is a cleavable linker utilized in the creation of antibody-drug conjugates (ADCs) and as a PEG-based PROTAC linker for the synthesis of PROTACs. It plays a role in preparing ADC precursors for specific cysteine mutant conjugation.
  • Functions as a cleavable linker.
  • Can be used to synthesize linker-payloads for conjugation.
  • Applicable in the preparation of ADCs.
  • Useful as a PEG-based linker for PROTAC synthesis.
  • Aids in creating neolymphostin-based ADC precursors.