Learn More
Medchemexpress LLC Maleimidoacetic acid N-hydroxysuccinimide ester | 55750-61-3 | MFCD00133523 | 99.6% | 252.18 | C10H8N2O6 | 25 G
SDP
Catalog No. 5000203539
Encompass_Preferred
AMAS (maleimidoacetic acid N-hydroxysuccinimide ester) is a noncleavable heterobifunctional crosslinker containing an NHS ester and a maleimide group that enables covalent conjugation between amine- and sulfhydryl-containing molecules for stable bioconjugation.
  • Noncleavable heterobifunctional crosslinker enabling stable covalent bonds
  • Contains NHS ester for amine coupling
  • Contains maleimide for thiol-specific conjugation
  • Suitable for antibody-drug conjugates and protein labeling
  • Available in multiple package sizes, including 25 G
Catalog No. 50-002-03539 Supplier Medchemexpress LLC Supplier No. HY12892525G
AMAS (maleimidoacetic acid N-hydroxysuccinimide ester) is a noncleavable heterobifunctional crosslinker containing an NHS ester and a maleimide group that enables covalent conjugation between amine- and sulfhydryl-containing molecules for stable bioconjugation.
  • Noncleavable heterobifunctional crosslinker enabling stable covalent bonds
  • Contains NHS ester for amine coupling
  • Contains maleimide for thiol-specific conjugation
  • Suitable for antibody-drug conjugates and protein labeling
  • Available in multiple package sizes, including 25 G