Learn More
Medchemexpress LLC Maleimidoacetic acid N-hydroxysuccinimide ester | 55750-61-3 | MFCD00133523 | 99.6% | 252.18 g/mol | C10H8N2O6 | 250 MG
SDP
Catalog No. 5000204156
Encompass_Preferred
AMAS (maleimidoacetic acid N-hydroxysuccinimide ester) is a noncleavable heterobifunctional crosslinker that carries an N-hydroxysuccinimide (NHS) ester and a maleimide group. It enables covalent conjugation between primary amines and free thiols to form stable amide and thioether bonds, and is used for bioconjugation and linker chemistry in antibody-drug conjugates and other crosslinking applications.
  • Noncleavable heterobifunctional crosslinker with NHS ester and maleimide groups.
  • Enables amine-to-thiol conjugation forming stable amide and thioether bonds.
  • High purity suitable for sensitive bioconjugation workflows.
  • Soluble in DMSO (about 100 mg/mL); ultrasonic assistance recommended.
  • Store powder at -20°C; solutions require low-temperature storage.
Catalog No. 50-002-04156 Supplier Medchemexpress LLC Supplier No. HY128925250MG
AMAS (maleimidoacetic acid N-hydroxysuccinimide ester) is a noncleavable heterobifunctional crosslinker that carries an N-hydroxysuccinimide (NHS) ester and a maleimide group. It enables covalent conjugation between primary amines and free thiols to form stable amide and thioether bonds, and is used for bioconjugation and linker chemistry in antibody-drug conjugates and other crosslinking applications.
  • Noncleavable heterobifunctional crosslinker with NHS ester and maleimide groups.
  • Enables amine-to-thiol conjugation forming stable amide and thioether bonds.
  • High purity suitable for sensitive bioconjugation workflows.
  • Soluble in DMSO (about 100 mg/mL); ultrasonic assistance recommended.
  • Store powder at -20°C; solutions require low-temperature storage.