Learn More
Cayman Chemical Maleimidoacetic Acid N-hydroxysuccinimide ester, Purity: ≥95%, CAS: 55750-61-3, 100 mg
SDP
Catalog No. 502967856
Encompass_Preferred
A crosslinking reagent that contains NHS ester- and maleimide-reactive groups at the opposite ends of a 4.4 Å spacer arm; used for sequential, two-stage conjugation with amine and sulfhydryl functional groups in the preparation of protein-hapten or protein-protein conjugates
Catalog No. 50-296-7856 Supplier Cayman Chemical Supplier No. 17160100 MG
A crosslinking reagent that contains NHS ester- and maleimide-reactive groups at the opposite ends of a 4.4 Å spacer arm; used for sequential, two-stage conjugation with amine and sulfhydryl functional groups in the preparation of protein-hapten or protein-protein conjugates