Learn More
AdipoGen MaleimidoEt-HBPO-pyridinium Tf
SDP
Catalog No. 502560536
Encompass_Preferred
Chemical. CAS 155863-05-1. Formula C24H20F3N3O7S. MW 551.49. Synthetic. Highly photostable, water soluble fluorescent label with large Stokes shift, high QY in aqueous media and pH tolerance.
Catalog No. 50-256-0536 Supplier AdipoGen Supplier No. CDXM0081M005
Chemical. CAS 155863-05-1. Formula C24H20F3N3O7S. MW 551.49. Synthetic. Highly photostable, water soluble fluorescent label with large Stokes shift, high QY in aqueous media and pH tolerance.