Learn More
AdipoGen MaleimidoEt-MePO-pyridinium Tf
SDP
Catalog No. 502560539
Encompass_Preferred
Chemical. CAS 155862-98-9. Formula C22H18F3N3O7S. MW 525.45. Synthetic. Highly photostable, water soluble fluorescent label with large Stokes shift, high QY in aqueous media and pH tolerance.
Catalog No. 50-256-0539 Supplier AdipoGen Supplier No. CDXM0080M005
Chemical. CAS 155862-98-9. Formula C22H18F3N3O7S. MW 525.45. Synthetic. Highly photostable, water soluble fluorescent label with large Stokes shift, high QY in aqueous media and pH tolerance.