Learn More
AdipoGen Malformin A1 (1 mg)
SDP
Supplier:  AdipoGen AGCN20169M001
Encompass_Preferred
Chemical. CAS: 3022-92-2. Formula: C23H39N5O5S2. MW: 529.7. Synthetic. Originally isolated from Aspergillus niger. Peptide antibiotic. Antibcaterial. Plant growth stimulator. Induces root curvature and malformation in plants. Mycotoxin. Prevents interleukin-1 (IL-1) induced endothelial changes by inhibition of protein synthesis. Inhibitor of interleukin-1 beta (IL1 beta) binding to its receptor. Enhancer of cellular fibrinolytic activity. Disrupts the cell cycle at the G2 checkpoint of cancer cells, leading to sensitization of the cancer cells to anti-cancer reagents. Anticancer compound. Cytotoxic against several cancer cell lines. Antimalarial and antitrypanosomal. Inhibitor of BRAF-mutated melanoma cell lines.
Catalog No. 50-205-3561