Learn More
AdipoGen Malformin C (250 µg)
SDP
Supplier:  AdipoGen AGCN20107C250
Encompass_Preferred
Chemical. CAS: 59926-78-2. Formula: C23H39N5O5S2. MW: 529.7. Synthetic. Originally isolated from Aspergillus niger FKI-2342. Peptide antibiotic. Induces root curvature and malformation in plants. Antibacterial, antimalarial and antitrypanosomal. Disrupts the cell cycle at the G2 checkpoint of cancer cells, leading to sensitization of the cancer cells to anti-cancer reagents.
Catalog No. 50-205-3466