Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malic Acid Standard, 0.3% (w/v), Ricca Chemical
SDP

Catalog No. R4581000250 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4581000250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4581000250 Supplier Ricca Chemical Company Supplier No. R4581000250A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97-67-6 , 7732-18-5
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2S)-2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.29
CAS Max % 100.0, 0.31
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 250 mL
Percent Purity 0.003
Grade Laboratory
Chemical Name or Material Malic Acid Standard, 0.3% w/v
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.