Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malic Acid Standard, 1500 ppm C4H6O5 in 10% (v/v) Ethanol, Ricca Chemical
SDP

Catalog No. R4583150100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4583150100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4583150100 Supplier Ricca Chemical Company Supplier No. R4583150100A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6915-15-7 , 64-17-5
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
InChI Key BJEPYKJPYRNKOW-UHFFFAOYSA-N
PubChem CID 525
ChEBI CHEBI:6650
IUPAC Name 2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Packaging Natural Poly Bottle
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Malic Acid Standard, 1500ppm in 10% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.