Learn More
Medchemexpress LLC Malic acid disodium hydrate | 63474-37-3 | 98.0% | 178.05 g/mol | C4H6O5·xH2O·2Na | 10 G
SDP
Catalog No. 5000281672
Encompass_Preferred
Malic acid disodium hydrate is the disodium salt of malic acid, supplied as a solid reagent used as a buffering agent and biochemical assay reagent. It dissolves readily in water and is commonly used to prepare buffers, control pH in assays, and as an acidity regulator in formulation work.
  • High water solubility for easy dissolution.
  • Useful for buffer preparation and pH control in biochemical assays.
  • Serves as an acidity regulator and ingredient in formulation studies.
  • Approximately 98.0% purity suitable for laboratory use.
  • Packaged for convenient handling and storage.
Catalog No. 50-002-81672 Supplier Medchemexpress LLC Supplier No. HYW250303A10G
Malic acid disodium hydrate is the disodium salt of malic acid, supplied as a solid reagent used as a buffering agent and biochemical assay reagent. It dissolves readily in water and is commonly used to prepare buffers, control pH in assays, and as an acidity regulator in formulation work.
  • High water solubility for easy dissolution.
  • Useful for buffer preparation and pH control in biochemical assays.
  • Serves as an acidity regulator and ingredient in formulation studies.
  • Approximately 98.0% purity suitable for laboratory use.
  • Packaged for convenient handling and storage.