Learn More
Medchemexpress LLC Malic acid disodium hydrate | 63474-37-3 | 98.0% | 178.05 g/mol | C4H6O5·xH2O·2Na | 25 G
SDP
Catalog No. 5000280881
Encompass_Preferred
Malic acid disodium hydrate is the disodium salt hydrate of malic acid used as a buffering agent and biochemical assay reagent. It is commonly used in buffer preparation, biochemical assays, and related research applications. Typical specifications include ≥98.0% purity, CAS 63474-37-3, formula C4H6O5·xH2O·2Na, and high water solubility when sonicated; store protected from moisture under nitrogen and follow recommended temperature guidelines for long-term stability.
  • High purity for reliable assay performance.
  • Readily soluble in water with ultrasonic assistance.
  • Suitable as a buffering agent in biochemical applications.
  • Hydrated form supports ease of handling and storage.
  • Stable under recommended storage and solvent conditions.
Catalog No. 50-002-80881 Supplier Medchemexpress LLC Supplier No. HYW250303A25G
Malic acid disodium hydrate is the disodium salt hydrate of malic acid used as a buffering agent and biochemical assay reagent. It is commonly used in buffer preparation, biochemical assays, and related research applications. Typical specifications include ≥98.0% purity, CAS 63474-37-3, formula C4H6O5·xH2O·2Na, and high water solubility when sonicated; store protected from moisture under nitrogen and follow recommended temperature guidelines for long-term stability.
  • High purity for reliable assay performance.
  • Readily soluble in water with ultrasonic assistance.
  • Suitable as a buffering agent in biochemical applications.
  • Hydrated form supports ease of handling and storage.
  • Stable under recommended storage and solvent conditions.