Learn More
Medchemexpress LLC Malic acid disodium hydrate | 63474-37-3 | 98.0% | 196.07 g/mol | C4H6O5·XH2O·2Na | 5 G
SDP
Catalog No. 5000281531
Encompass_Preferred
Malic acid disodium hydrate is the disodium salt hydrate form of malic acid intended for laboratory biochemical and buffering applications. It is a white to off-white powder with high water solubility and is supplied in small packs for laboratory use. The reagent is documented with storage and solubility data suited for assay workflows.
  • Disodium salt hydrate form of malic acid, useful as a buffer and biochemical reagent.
  • High purity: 98.0% suitable for analytical and assay work.
  • Water soluble: approximately 175 mg/mL (may require ultrasonic assistance).
  • Stable at room temperature when stored under nitrogen and away from moisture; extended storage in solvent at -20°C to -80°C.
  • Supplied in small laboratory pack sizes for convenient handling.
Catalog No. 50-002-81531 Supplier Medchemexpress LLC Supplier No. HYW250303A5G
Malic acid disodium hydrate is the disodium salt hydrate form of malic acid intended for laboratory biochemical and buffering applications. It is a white to off-white powder with high water solubility and is supplied in small packs for laboratory use. The reagent is documented with storage and solubility data suited for assay workflows.
  • Disodium salt hydrate form of malic acid, useful as a buffer and biochemical reagent.
  • High purity: 98.0% suitable for analytical and assay work.
  • Water soluble: approximately 175 mg/mL (may require ultrasonic assistance).
  • Stable at room temperature when stored under nitrogen and away from moisture; extended storage in solvent at -20°C to -80°C.
  • Supplied in small laboratory pack sizes for convenient handling.