Learn More
Medchemexpress LLC Malic acid disodium hydrate | 63474-37-3 | 98.0% | 196.07 g/mol | C4H6O5 · xH2O · 2Na | 500 MG
SDP
Catalog No. 5000280628
Encompass_Preferred
Malic acid disodium hydrate is the disodium salt/hydrate form of malic acid used as a biochemical reagent, buffering agent, and additive for nutritional and assay applications. Typical purity is 98.0% and it is supplied in small pack sizes for laboratory use.
  • High purity: 98.0% for reliable analytical use.
  • Buffered salt form suitable for pH control in biochemical assays.
  • Available in small packs (500 mg) for method development and testing.
  • Water-soluble, white crystalline powder for easy formulation.
  • Includes datasheet and SDS for safe handling and storage.
Catalog No. 50-002-80628 Supplier Medchemexpress LLC Supplier No. HYW250303A500MG
Malic acid disodium hydrate is the disodium salt/hydrate form of malic acid used as a biochemical reagent, buffering agent, and additive for nutritional and assay applications. Typical purity is 98.0% and it is supplied in small pack sizes for laboratory use.
  • High purity: 98.0% for reliable analytical use.
  • Buffered salt form suitable for pH control in biochemical assays.
  • Available in small packs (500 mg) for method development and testing.
  • Water-soluble, white crystalline powder for easy formulation.
  • Includes datasheet and SDS for safe handling and storage.