Learn More
Medchemexpress LLC Malic acid disodium salt | 676-46-0 | MFCD00012466 | 98.0% | 178.05 | C4H4Na2O5 | 25 G
SDP
Catalog No. 5000281391
Encompass_Preferred
Malic acid disodium salt is the disodium salt of malic acid (sodium DL-malate) supplied as a high-purity biochemical reagent. It is commonly used as a buffering agent, pH regulator, and reagent in biochemical assays and food additive research.
  • High purity (98.0%) for reliable experimental results.
  • Useful as a buffering agent in aqueous solutions.
  • Suitable for biochemical assay and buffer preparation.
  • White crystalline powder, easy to weigh and handle.
  • Molecular formula C4H4Na2O5; molecular weight 178.05.
Catalog No. 50-002-81391 Supplier Medchemexpress LLC Supplier No. HYW25030325G
Malic acid disodium salt is the disodium salt of malic acid (sodium DL-malate) supplied as a high-purity biochemical reagent. It is commonly used as a buffering agent, pH regulator, and reagent in biochemical assays and food additive research.
  • High purity (98.0%) for reliable experimental results.
  • Useful as a buffering agent in aqueous solutions.
  • Suitable for biochemical assay and buffer preparation.
  • White crystalline powder, easy to weigh and handle.
  • Molecular formula C4H4Na2O5; molecular weight 178.05.