Learn More
Medchemexpress LLC Malic acid disodium salt | 676-46-0 | MFCD00012466 | 98.0% | 178.05 g/mol | C4H4Na2O5 | 10 G
SDP
Catalog No. 5000280615
Encompass_Preferred
Malic acid disodium salt is the disodium salt of malic acid, provided as a white crystalline powder suitable for use as a buffering agent, pH regulator, food additive, and biochemical reagent for in vitro research and analytical applications.
  • White crystalline powder, soluble in water.
  • Function as a buffering agent and pH regulator in biochemical assays.
  • Useful as a food additive and nutritional supplement ingredient.
  • Molecular formula C4H4Na2O5; molecular weight 178.05 g/mol; CAS 676-46-0.
  • Typical purity 98.0% as supplied for laboratory use.
  • Available in small laboratory pack sizes for analytical workflows.
Catalog No. 50-002-80615 Supplier Medchemexpress LLC Supplier No. HYW25030310G
Malic acid disodium salt is the disodium salt of malic acid, provided as a white crystalline powder suitable for use as a buffering agent, pH regulator, food additive, and biochemical reagent for in vitro research and analytical applications.
  • White crystalline powder, soluble in water.
  • Function as a buffering agent and pH regulator in biochemical assays.
  • Useful as a food additive and nutritional supplement ingredient.
  • Molecular formula C4H4Na2O5; molecular weight 178.05 g/mol; CAS 676-46-0.
  • Typical purity 98.0% as supplied for laboratory use.
  • Available in small laboratory pack sizes for analytical workflows.