Learn More
Medchemexpress LLC Malic acid disodium salt | 676-46-0 | MFCD00012466 | 98.0% | 178.05 g/mol | C4H4Na2O5 | 5 G
SDP
Catalog No. 5000281530
Encompass_Preferred
Malic acid disodium salt is the disodium salt form of malic acid supplied as a white crystalline powder for laboratory and research use. It is commonly used as a buffering agent and reagent in biochemical assays, food additive research, and buffer preparations. The substance has CAS 676-46-0, molecular formula C4H4Na2O5, molecular weight 178.05 g/mol, and typical purity of 98.0%.
  • Suitable for biochemical assays and buffer preparation.
  • High purity (98.0%) for reproducible analytical results.
  • White crystalline solid that dissolves easily for solution preparation.
  • Applicable in food additive and formulation research (E 296 disodium salt).
  • Available in small laboratory pack sizes for convenient handling.
Catalog No. 50-002-81530 Supplier Medchemexpress LLC Supplier No. HYW2503035G
Malic acid disodium salt is the disodium salt form of malic acid supplied as a white crystalline powder for laboratory and research use. It is commonly used as a buffering agent and reagent in biochemical assays, food additive research, and buffer preparations. The substance has CAS 676-46-0, molecular formula C4H4Na2O5, molecular weight 178.05 g/mol, and typical purity of 98.0%.
  • Suitable for biochemical assays and buffer preparation.
  • High purity (98.0%) for reproducible analytical results.
  • White crystalline solid that dissolves easily for solution preparation.
  • Applicable in food additive and formulation research (E 296 disodium salt).
  • Available in small laboratory pack sizes for convenient handling.