Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Malic acid disodium salt | 676-46-0 | MFCD00012466 | 98.0% | 178.05 g/mol | C4H4Na2O5 | 500 MG
SDP

Catalog No. 5000281392
Encompass_Preferred

Malic acid disodium salt (sodium DL-malate) is the disodium salt of malic acid used as a buffering agent and biochemical reagent. It is commonly used to adjust solution acidity, act as a food additive or nutritional supplement, and support metabolic or biochemical assays.

  • High purity: 98.0%.
  • Molecular weight: 178.05 g/mol.
  • CAS number: 676-46-0.
  • Useful as a buffering agent and reagent in biochemical assays.
  • Suitable for use as a food additive or nutritional supplement.
  • Available in a 500 mg pack size.

Catalog No. 50-002-81392 Supplier Medchemexpress LLC Supplier No. HYW250303500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Malic acid disodium salt (sodium DL-malate) is the disodium salt of malic acid used as a buffering agent and biochemical reagent. It is commonly used to adjust solution acidity, act as a food additive or nutritional supplement, and support metabolic or biochemical assays.

  • High purity: 98.0%.
  • Molecular weight: 178.05 g/mol.
  • CAS number: 676-46-0.
  • Useful as a buffering agent and reagent in biochemical assays.
  • Suitable for use as a food additive or nutritional supplement.
  • Available in a 500 mg pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.