Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malic Acid, NF, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604154 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-154 500 g Poly Bottle
18-604-155 2.5 kg Poly Pail
18-604-156 12 kg Poly Pail
18-604-157 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-154 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1336500GM
Add to Cart
Add to Cart

M1336, 617-48-1, C4H6O5

Spectrum™ Chemical Malic Acid, NF is used as a pH modifier in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 617-48-1
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
InChI Key BJEPYKJPYRNKOW-UHFFFAOYNA-N
IUPAC Name 2-hydroxybutanedioic acid
SMILES OC(CC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Percent Purity 99 to 100.5%
Grade NF
Quantity 500 g
Ignition Residue 0.1%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.