Learn More
Sigma Aldrich Fine Chemicals Biosciences Malic Dehydrogenase from p
Catalog No. 502900209
Encompass
Malic dehydrogenase is a cytoplasmic isozyme and an important catalyst in the tricarboxylic acid cycle.
Catalog No. 50-290-0209 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M156710KU
Malic dehydrogenase is a cytoplasmic isozyme and an important catalyst in the tricarboxylic acid cycle.