Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Malic dehydrogenase, microorganism (MDH) | 9001-64-3 | ≥10 U/mg solid | 35,000 | 5 KU
SDP

Catalog No. 5000397435
Encompass_Preferred

Malate dehydrogenase (EC 1.1.1.37) (MDH) catalyzes the mutual conversion of oxaloacetate and malate, and is associated with the oxidation/reduction of dinucleotide coenzymes.

  • Michaelis constant: 1.5x10-6 M (NADH, 37°C), 1.9x10-6 M (oxaloacetate, 37°C)
  • Optimum pH: 7.5-8.0
  • Optimum temperature: 60°C
  • PH stability: 3.0-9.0 (37°C, 30 minutes)
  • Thermal stability: below 65°C (pH 7.5, 30 minutes)
  • Enzyme activity: ≥10 U/mg solid
  • Unit definition: one unit is defined as the amount of enzyme that oxidation of 1 μmol NADH per min at pH 7.5 at 30°C.
  • Appearance: solid
  • Color: white to off-white
  • EC number: 1.1.1.37
  • Solubility: H2O : ≥ 100 mg/mL

Catalog No. 50-003-97435 Supplier Medchemexpress LLC Supplier No. HYP28095KU
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Malate dehydrogenase (EC 1.1.1.37) (MDH) catalyzes the mutual conversion of oxaloacetate and malate, and is associated with the oxidation/reduction of dinucleotide coenzymes.

  • Michaelis constant: 1.5x10-6 M (NADH, 37°C), 1.9x10-6 M (oxaloacetate, 37°C)
  • Optimum pH: 7.5-8.0
  • Optimum temperature: 60°C
  • PH stability: 3.0-9.0 (37°C, 30 minutes)
  • Thermal stability: below 65°C (pH 7.5, 30 minutes)
  • Enzyme activity: ≥10 U/mg solid
  • Unit definition: one unit is defined as the amount of enzyme that oxidation of 1 μmol NADH per min at pH 7.5 at 30°C.
  • Appearance: solid
  • Color: white to off-white
  • EC number: 1.1.1.37
  • Solubility: H2O : ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.