Learn More
Medchemexpress LLC Malic enzyme inhibitor ME1 | 522649-59-8 | 99.7% | 351.40 g·mol⁻¹ | C20H21N3O3 | 50MG
SDP
Catalog No. 5000235970
Encompass_Preferred
Malic enzyme inhibitor ME1 is a small-molecule, selective inhibitor of cytosolic malic enzyme (ME1) used for biochemical and cellular research. It exhibits potent inhibition of ME1 (IC50 = 0.15 μM) and is supplied as a white to off-white solid with high purity and defined storage recommendations.
  • Potent ME1 inhibition (IC50 = 0.15 μM).
  • High purity suitable for biochemical assays (99.7%).
  • Supplied as a solid, white to off-white.
  • Highly soluble in DMSO; may require sonication and warming.
  • Stable when stored under recommended conditions in powder or solution.
Catalog No. 50-002-35970 Supplier Medchemexpress LLC Supplier No. HY12486150MG
Malic enzyme inhibitor ME1 is a small-molecule, selective inhibitor of cytosolic malic enzyme (ME1) used for biochemical and cellular research. It exhibits potent inhibition of ME1 (IC50 = 0.15 μM) and is supplied as a white to off-white solid with high purity and defined storage recommendations.
  • Potent ME1 inhibition (IC50 = 0.15 μM).
  • High purity suitable for biochemical assays (99.7%).
  • Supplied as a solid, white to off-white.
  • Highly soluble in DMSO; may require sonication and warming.
  • Stable when stored under recommended conditions in powder or solution.