Learn More
Medchemexpress LLC Malic enzyme inhibitor ME1 | 522649-59-8 | 99.7% | 351.40 | 1 MG
SDP
Catalog No. 5000365345
Encompass_Preferred
Malic enzyme inhibitor ME1 is a potent inhibitor of Malic enzyme (ME1) with an IC50 of 0.15 μM. It is intended for research use only and has been shown to significantly impact cell viability and proliferation in various cell lines.
  • Potent inhibitor of Malic enzyme (ME1) with an IC50 of 0.15 μM
  • Significantly reduces total cell numbers and cell viability/metabolic activity in MTS cells
  • Dose-dependently reduces the number of colonies formed by HCT116 and HT29 cells
  • Suitable for research applications
Catalog No. 50-003-65345 Supplier Medchemexpress LLC Supplier No. 1MGHY124861
Malic enzyme inhibitor ME1 is a potent inhibitor of Malic enzyme (ME1) with an IC50 of 0.15 μM. It is intended for research use only and has been shown to significantly impact cell viability and proliferation in various cell lines.
  • Potent inhibitor of Malic enzyme (ME1) with an IC50 of 0.15 μM
  • Significantly reduces total cell numbers and cell viability/metabolic activity in MTS cells
  • Dose-dependently reduces the number of colonies formed by HCT116 and HT29 cells
  • Suitable for research applications