Learn More
Medchemexpress LLC HY-124861 5mg Medchemexpress, Malic enzyme inhibitor ME1 CAS:522649-59-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1248615MG
Encompass_Preferred
Medchemexpress, HY-124861 5mg Malic enzyme inhibitor ME1 CAS:522649-59-8 Malic enzyme inhibitor ME1 (ME1; compound 1) is a potent inhibitor of Malic enzyme (ME1) with an IC50 of 0.15 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9683