Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Mall Probe 6 in.

Catalog No. 13820026 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
13-820-026 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-820-026
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fisherbrand™ Mall Probe 6 in. Nickel-Plated Steel Probe is equipped with a round handle that is partly knurled to ensure a firm grip.

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Product Type Probe
Autoclavable Autoclavable
Color Silver
For Use With (Application) Dissecting applicatons
Includes Probe
Latex Free Yes
Quantity 1
Size 6 in.
Length (English) 6 in.
Length (Metric) 152.4 mm
Width (Metric) N/A
Height (Metric) N/A
Weight (Metric) N/A
Electrical Requirements N/A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.