Learn More
Kingfisher Biotech Inc Human CXCL9 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY2048MD004
Encompass_Preferred
The Human CXCL9 (MIG) ELISA contains capture antibody, standard, and detection antibody for development of a Human CXCL9 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Human CXCL9 ELISA can also be used to measure Chimpanzee CXCL9. Alternate names include CXCL9, CMK, Humig, MIG, SCYB9, crg-10, C-X-C motif chemokine ligand 9. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5662