Learn More
Braintree Scientific Malleable S/S Disposable Feeding Needle,Sterile, 18G-2""
SDP
Catalog No. 501954609
Encompass
Malleable S/S Disposable Feeding Needle. These needles feature plastic luer hubs and silicone tips and can be easily formed to desired shape. 100 sterile needles per box. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4609 Supplier Braintree Scientific Supplier No. D002
 Restricted Item
Add to Cart
Add to Cart
Malleable S/S Disposable Feeding Needle. These needles feature plastic luer hubs and silicone tips and can be easily formed to desired shape. 100 sterile needles per box. **Sold for research purposes only and not for human use**