Learn More
Bioworld Malmgren Terrestrial Orchid Medium, 25 L
Supplier:  Bioworld 306301852
Encompass_Preferred
Malmgren Terrestrial Orchid MediumContains the Macronutrients, Micronutrients, and Organic Constituents as described by Malmgren (1996)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.84 g/LPartially soluble in water
Catalog No. 50-254-958