Learn More
Bioworld Malmgren Terrestrial Orchid Medium, w/ Pineapple Powder, 50 L
Supplier:  Bioworld 306301864
Encompass_Preferred
Malmgren Modified Terrestrial Orchid MediumContains the Macronutrients, Micronutrients, and Organic Constituents as described by Malmgren (1996) with addition of Pineapple PowderSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 21.84 g/LPartially soluble in water
Catalog No. 50-254-962