Learn More
Cayman Chemical MalntIdetrIfluoroacetAT s 10mg
SDP
Catalog No. 502942471
Encompass_Preferred
A fluorescent substrate for PKA and PKC; upon phosphorylation by PKA or PKC, the intrinsic tyrosine fluorescence decreases and can be used to quantify PKA and PKC activity; ex/em = 285/302 nm
Catalog No. 50-294-2471 Supplier Cayman Chemical Supplier No. 4015010 MG
A fluorescent substrate for PKA and PKC; upon phosphorylation by PKA or PKC, the intrinsic tyrosine fluorescence decreases and can be used to quantify PKA and PKC activity; ex/em = 285/302 nm