Learn More
Cayman Chemical MaLNyl Coenzyme AlIthIM1mg
SDP
Catalog No. 502969858
Encompass_Preferred
A CoA derivative used in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane; may mediate cancer cell cytotoxicity induced by fatty acid synthase inhibition
Catalog No. 50-296-9858 Supplier Cayman Chemical Supplier No. 164551 MG
A CoA derivative used in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane; may mediate cancer cell cytotoxicity induced by fatty acid synthase inhibition