Learn More
Cayman Chemical MaLNyl Coenzyme AlIthIM25mg
SDP
Catalog No. 502970949
Encompass_Preferred
A CoA derivative used in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane; may mediate cancer cell cytotoxicity induced by fatty acid synthase inhibition
Catalog No. 50-297-0949 Supplier Cayman Chemical Supplier No. 1645525 MG
A CoA derivative used in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane; may mediate cancer cell cytotoxicity induced by fatty acid synthase inhibition