Learn More
Cayman Chemical MaLNyl Coenzyme AlIthIM5mg
SDP
Catalog No. 502970950
Encompass_Preferred
A CoA derivative used in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane; may mediate cancer cell cytotoxicity induced by fatty acid synthase inhibition
Catalog No. 50-297-0950 Supplier Cayman Chemical Supplier No. 164555 MG
A CoA derivative used in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane; may mediate cancer cell cytotoxicity induced by fatty acid synthase inhibition