Learn More
Cayman Chemical MalolactomycIn C 100ug
SDP
Catalog No. 502991259
Encompass_Preferred
A bacterial metabolite; active against B. cinerea, C. fulvum, and P. oryzae (MIC = 25 UG/ml for all)
Catalog No. 50-299-1259 Supplier Cayman Chemical Supplier No. 31673100 UG
A bacterial metabolite; active against B. cinerea, C. fulvum, and P. oryzae (MIC = 25 UG/ml for all)