Learn More
AdipoGen anti-Malondialdehyde [MDA], mAb (1F83) (30 µg)
SDP
Supplier:  AdipoGen JAIMMD030N
Encompass_Preferred
anti-Malondialdehyde [MDA], mAb (1F83) (30 µg), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes MDA-modified proteins (especially DHP-lysine). Frozen liquid. Contains 10mM PBS, 0.1% NaN3 and 0.5% BSA. Malondialdehyde (MDA) is one of the major aldehyde derive from lipid peroxidation. MDA is highly reactive aldehyde and reacts with lysine residue in protein. The reaction with MDA and lysine residue leads to the formation of numerous numbers of adducts, such as dihydropyridine-lysine (DHP-lysine) type derivative.
Catalog No. 50-148-9751