Learn More
Enzo Life Sciences Malondialdehyde polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210879R100
Encompass_Preferred
Host: Rabbit. Immunogen: Malondialdehyde (MDA) conjugated to KLH. Species reactivity: Species independent. Specificity: Recognizes MDA adducts. Applications: ELISA, IHC, WB. Recommended Dilutions: ELISA (1:1,000-1:10,000) Immunohistochemistry (1:500-1:1,000) Western blot (1:500-1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Unpurified. Formulation: Liquid. Contains 0.05% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8179