Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Malondialdehyde Tetrabutylammonium Salt
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Malondialdehyde Tetrabutylammonium Salt
Spécifications
Spécifications
| CAS | 100683-54-3 |
| Melting Point | 126°C to 131°C |
| Molecular Formula | C19H39NO2 |
| Linear Formula | OCHCH=CHO[N(CH2CH2CH2CH3)4] |
| MDL Number | MFCD00043410 |
| Quantity | 1 g |
| Synonym | Malonaldehyde tetrabutylammonium salt; Tetrabutylammonium malondialdehyde enolate |
| Molecular Weight (g/mol) | 313.52 g/mol |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'