Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Malondialdehyde Tetrabutylammonium Salt

Catalog No. 632875GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-287-5GF 5 g
63-287-1GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-287-5GF Supplier Sigma Aldrich Supplier No. 632875GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Malondialdehyde Tetrabutylammonium Salt

Specifications

CAS 100683-54-3
Melting Point 126°C to 131°C
Molecular Formula C19H39NO2
Linear Formula OCHCH=CHO[N(CH2CH2CH2CH3)4]
MDL Number MFCD00043410
Quantity 5 g
Synonym Malonaldehyde tetrabutylammonium salt; Tetrabutylammonium malondialdehyde enolate
Molecular Weight (g/mol) 313.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.