Learn More
Apexbio Technology LLC Malonic acid 141-82-2 500mg
SDP
Catalog No. 5000087996
Encompass_Preferred
Malonic acid is a small-molecule dicarboxylic acid It is commonly used to facilitate the cross-linking of polysaccharide molecules such as corn starch and potato starch by forming covalent linkages via its two carboxyl groups Malonic acid acts primarily through condensation or esterification reactions to increase intermolecular connectivity and modulate the mechanical properties of biopolymer matrices Based on these properties malonic acid holds research potential in pharmaceutical excipient formulation drug delivery system development and biomaterial engineering studies
Catalog No. 50-000-87996 Supplier Apexbio Technology LLC Supplier No. B7900500
Malonic acid is a small-molecule dicarboxylic acid It is commonly used to facilitate the cross-linking of polysaccharide molecules such as corn starch and potato starch by forming covalent linkages via its two carboxyl groups Malonic acid acts primarily through condensation or esterification reactions to increase intermolecular connectivity and modulate the mechanical properties of biopolymer matrices Based on these properties malonic acid holds research potential in pharmaceutical excipient formulation drug delivery system development and biomaterial engineering studies