Learn More
TARGETMOL CHEMICALS INC MALONIC ACID 500MG
SDP
Catalog No. 502576694
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Malonic acid (propanedioic acid) is the archetypal example of a competitive inhibitor it acts against succinate dehydrogenase (complex II) in the respiratory electron transport chain. Malonic acid is found to be associated with malonyl-CoA decarboxylase deficiency which is an inborn error of metabolism. purity: 98%
Catalog No. 50-257-6694 Supplier TARGETMOL CHEMICALS INC Supplier No. T5291500MG
Also available in 25 mg 50 mg 100 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Malonic acid (propanedioic acid) is the archetypal example of a competitive inhibitor it acts against succinate dehydrogenase (complex II) in the respiratory electron transport chain. Malonic acid is found to be associated with malonyl-CoA decarboxylase deficiency which is an inborn error of metabolism. purity: 98%